missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46811
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Polyclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TWEAK/TNFSF12 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| O43508 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QEPAQEELVAEEDQDPSELNPQTEESQDPAPFLNRLVRPRRSAPKGRKTRARRAIAAHYEVHPRPGQDGAQAGVDGTVSGWEEARINSSSPLRYNRQIGEFIVTRAGLYYLYC | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| 8742 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction