Learn More
Fisher Chemical 250 ML Twenty three elements solution - Ag, Al, As, Ba, Be, Bi, Cd, Co, Cu, Cr, Fe, Ga, In, Ni, each 2% HNO3 matrix 250mL
Brand: Fisher Chemical CM1728
250 ML Twenty three elements solution - Ag, Al, As, Ba, Be, Bi, Cd, Co, Cu, Cr, Fe, Ga, In, Ni, each 2% HNO3 matrix 250mL
Additional Details : Weight : 0.25000kg