missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWF1 Monoclonal Antibody (OTI5C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808892
This item is not returnable.
View return policy
Description
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.Specifications
| TWF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12792 | |
| TWF1 | |
| Human recombit protein fragment corresponding to amino acids 140-384 of human TWF1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5C11 | |
| Unconjugated | |
| TWF1 | |
| A6, PTK9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5756 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction