missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWF2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94579-0.1ml
This item is not returnable.
View return policy
Description
TWF2 Polyclonal antibody specifically detects TWF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TWF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunoprecipitation 1:20-1:50 | |
| A6-related protein, A6RPFLJ56277, hA6RP, Protein tyrosine kinase 9-like, protein tyrosine kinase 9-like (A6-related protein), PTK9L protein tyrosine kinase 9-like (A6-related protein), PTK9LA6r, twinfilin, actin-binding protein, homolog 2 (Drosophila), Twinfilin-1-like protein, twinfilin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human TWF2 (NP_009215.1). MAHQTGIHATEELKEFFAKARAGSVRLIKVVIEDEQLVLGASQEPVGRWDQDYDRAVLPLLDAQQPCYLLYRLDSQNAQGFEWLFLAWSPDNSPVRLKMLYAATRATVKKEFGGGHIKDELFGTVKDDLSFAGYQKHLSSCAAPAPLTSAERELQQIRINEVKTEISVESKHQTLQGLAFPLQPEAQRALQQLKQKMVNYIQMKLDLERETIELVHTEPTDVAQLPSRVPRDAARYHFFLYKHTHEGDPLESVVFIYSMPGYKCSIKERMLYSSCKSRLLDSVEQDFHLEIAKKIEIGDGAELTAEFLYDEVHPKQHAFKQAFAKPKGPGGKRGHKRLIRGPGENGDDS | |
| 0.1 mL | |
| Protein Kinase | |
| 11344 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction