missing translation for 'onlineSavingsMsg'
Learn More

TWF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 190.00 - € 550.00

Specifications

Antigen TWF2
Dilution ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30227442
View Documents
Novus Biologicals
NBP3-37927-20ul
20 μL
€ 190.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30230417
View Documents
Novus Biologicals
NBP3-37927-100ul
100 μL
€ 550.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TWF2 Polyclonal antibody specifically detects TWF2 in Human samples. It is validated for ELISA,Immunoprecipitation
Specifications

Specifications

TWF2
ELISA, Immunoprecipitation
Unconjugated
Rabbit
Protein Kinase
PBS (pH 7.3), 50% glycerol
11344
IgG
Affinity purified
ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells
Polyclonal
Purified
RUO
Human
A6-related protein, A6RPFLJ56277, hA6RP, Protein tyrosine kinase 9-like, protein tyrosine kinase 9-like (A6-related protein), PTK9L protein tyrosine kinase 9-like (A6-related protein), PTK9LA6r, twinfilin, actin-binding protein, homolog 2 (Drosophila), Twinfilin-1-like protein, twinfilin-2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-369 of human TWF2 (NP_009215.1).,, Sequence:, MAHQTGIHATEELKEFFAKARAGSVRLIKVVIEDEQLVLGASQEPVGRWDQDYDRAVLPLLDAQQPCYLLYRLDSQNAQGFEWLFLAWSPDNSPVRLKMLYAATRATVKKEFGGGHIKDELFGTVKDDLSFAGYQKHLSSCAAPAPLTSAERELQQIRINEVKTEISVESKHQTLQGLAFPLQPEAQRALQQLKQKMVNYIQMKLDLERETIELVHTEPTDVAQLPSRVPRDAARYHFFLYKHTHEGDPLESVVFIYSMPGYKCSIKERMLYSSCKSRLLDSVEQDFHLEIAKKIEIGDGAELTAEFLYDEVHPKQHAFKQAFAKPKGPGGKRGHKRLIRGPGENGDDS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.