missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TWF2 Monoclonal Antibody (2B5)
GREENER_CHOICE

Product Code. 15682275
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15682275 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15682275 Supplier Invitrogen™ Supplier No. MA524450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: FAGYQKHLSS CAAPAPLTSA ERELQQIRIN EVKTEISVES KHQTLQGLAF PLQPEAQRAL QQLKQKMVNY IQMKLDLERE TIELVHTEPT DVAQLPSRVP.

TWF2 encodes a protein that was identified by its interaction with the catalytic domain of protein kinase C-zeta. The encoded protein contains an actin-binding site and an ATP-binding site. It is most closely related to twinfilin, a conserved actin monomer-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWF2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B5
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TWF2
Gene Accession No. Q6IBS0
Gene Alias A6 related protein; A6r; A6-related; A6-related protein; A6RP; AU014993; hA6RP; mA6RP; MSTP011; protein tyrosine kinase 9 related protein; Protein tyrosine kinase 9-like; protein tyrosine kinase 9-like (A6-related protein); PTK 9L; PTK9 protein typrotein tyrosine kinase 9-like (A6-related protein); Ptk9l; PTK9L protein tyrosine kinase 9-like (A6-related protein); Ptk9r; TWF 2; Twf2; Twinfilin 2; twinfilin actin binding protein 2; twinfilin actin-binding protein 2; twinfilin, actin-binding protein, homolog 2; twinfilin, actin-binding protein, homolog 2 (Drosophila); twinfilin-1-like protein; Twinfilin2; Twinfilin-2
Gene Symbols TWF2
Host Species Mouse
Immunogen PTK9L (AAH00327, 131 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11344
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.