Learn More
Julabo™ Twin Distributing Adapter
Product Code. 15264303
Click to view available options
Connection Type:
1x M24 x 1.5 F to 2x M24 x 1.5 M
1x M30 x 1.5 F to 2x M30 x 1.5 M
1x M38 x 1.5 F to 2x M38 x 1.5 M
Unit Size:
Each
Product Code. 15264303
Brand: Julabo™ 8890140
Only null left
Add to basket
Add to basket
Specifications
For Use With (Equipment) Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
Type Twin Distributing Adapter
Connection Type 1x M24 x 1.5 F to 2x M24 x 1.5 M