Learn More
IBA Lifesciences Twin-Strep-tag™ Capture Maxi Kit
Used for reversible capture of Twin-Strep-tag™ fusion proteins for SPR analyses
Brand: IBA Lifesciences 2-4370-010
Additional Details : Weight : 0.30800kg
Description
- Twin-Strep-tag™ Capture Kit is intended for site-directed, reversible capture of Twin-Strep-tag™ fusion proteins for biomolecular interaction analysis using surface plasmon resonance (SPR) devices
- The kit provides Strep-Tactin™XT, SPR Immobilization Buffer, Regeneration Buffer, and GFP-Twin-Strep-tag™ as control protein. It enables the highly stable immobilization of the ligand based on the unique Strep-Tactin™XT:Twin-Strep-tag™ interaction, which operates with pM affinity
- The key benefits are: Indirect immobilization protecting the ligand from chemical modification as well as inactivation or manipulation by the immobilization procedure. High affinity enabling measurements over a long period (high affinity in low pM range; T1/2 = 13 h). Chip can be regenerated easily by an established procedure – cost effective. Ligands can be captured directly from complex samples due to the high affinity and specificity of the Twin-Strep-tag™:Strep-Tactin™XT interaction
- The use of unconjugated Strep-Tactin™XT requires a license that is provided with the purchase of the kit as limited use license.
Specifications
Maxi | |
Store all kit components at 2°C to 8°C and GFP-Twin-Strep-tag at -20°C aftersolubilization. | |
Lyophilized |
SPR Immobilization Buffer, Regeneration Buffer, Strep-Tactin™XT , GFP-Twin-Strep-tag | |
SPR Device | |
Biomolecular Interaction Analysis Using Surface Plasmon Resonance |
Safety and Handling
- Regeneration buffer SPR-chips
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride
For Research Use Only.