Learn More
Manutan Twine, titration: 6/3 average, wire length: 500 m, wire Ø: 3.5 mm
Brand:  Manutan A016036
Encompass_Preferred
Twine, titration: 6/3 average, wire length: 500 m, wire Ø: 3.5 mm
Product Code. 15506498
  • € 32.07 / Pack of 1
Only null left
Estimated Shipment: 04-06-2026