Learn More
LABPLAS Twirl'Em Clean Room Writing
Product Code. 17920295
Click to view available options
Dimensions (L x W):
114 x 305 mm
140 x 229 mm
140 x 381 mm
178 x 305 mm
Unit Size:
Pack of 1000
Product Code. 17920295
Brand: LABPLAS EPR5515S
Only null left
Add to basket
Add to basket
Sterile sample bags that are are safe, flexible, contaminant-free, easy to use and transparent to ensure full visibility of your sample.
  • Twirl'Em clean room sampling bags
  • Safety tabs
  • Clear, with writing area for easy labeling
  • Closure with 1 round wire and 1 flat wire
  • Available in various sizes
  • Option between heavy or standard closure
Specifications
Autoclavable Not Autoclavable
Color Clear with White Block
Disposable Single-use
Sterility Sterile
Thickness (Metric) 76.2 μm
Latex Free Yes
Thickness (English) 3 mil.
Dimensions (L x W) 140 x 381 mm
Length (English) 5.5 in.
Length (Metric) 140 mm
Width (English) 15 in.
Width (Metric) 381 mm
Product Type Sample Bag