missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWIST2 Monoclonal Antibody (OTI6H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805212S
This item is not returnable.
View return policy
Description
TWIST2 Monoclonal antibody specifically detects TWIST2 in Human samples. It is validated for Western BlotSpecifications
| TWIST2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48307 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7980 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6H10 | |
| Unconjugated | |
| TFPI2 | |
| AMS; BBRSAY; bHLHa39; Class A basic helix-loop-helix protein 39; dermis expressed 1; dermis-expressed protein 1; DERMO1; Dermo-1; dermo-1 protein; FFDD3; SETLSS; twist basic helix-loop-helix transcription factor 2; twist family bHLH transcription factor 2; twist homolog 2; TWIST2; twist-related bHLH protein Dermo1; twist-related protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 23-235 of human TFPI2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction