Learn More
Köttermann Two Door Tall Storage Cabinet
This Two Door Tall Storage Cabinet features a two hinged or sliding doors to protect stored equipment and utensils.
Brand: Köttermann 301-00017
Additional Details : Weight : 119.29510kg
Description
- Tall, slender design expands storage with a small footprint
- Two doors protect stored items
- Sliding and hinged door options available
Specifications
None | |
Storage | |
No | |
119.3 kg | |
4 | |
Epoxy Coated | |
White |
None | |
No | |
500 x 1200 x 1920 mm | |
Sliding | |
Tall Storage | |
2 | |
Steel |