Learn More
Hamilton™ Custom Two-Hub Assembled Tubing Assemblies
Product Code. 10346302
Change view
Click to view available options
Quantity:
1/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10346302 1/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10346302 Supplier Hamilton™ Supplier No. 86510
Only null left
Add to basket
Add to basket
Chemically inert for fluid handling in the laboratory.
  • Custom length and gauge
  • Female luer fitting
  • Tubing manufactured from PTFE
Specifications
Type Tube
Quantity 1/Pk.
Material PTFE
Length (Metric) Custom
Needle Gauge Custom Gauge