Learn More
Thermo Scientific™ TX-750 Swinging Bucket/BIOShield 1000A Adapters
Product Code. 11894471 Shop All Thermo Scientific Products
Click to view available options
Capacity:
1.5/2.0 mL Conical Microtube
10 mL Blood Collection/15mL DIN Tube
100 mL Round-bottom Open-top Tube
14 mL Flanged Round-bottom Tube
15 mL Conical Urine Tube
150 mL Round-bottom Open-top Tube
25 mL DIN Round-bottom Tube
25 mL Universal Tube
250 mL Flat-bottom Bottles
45 mL Flat/Round Tube
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube
50 mL Round-bottom Tube
Unit Size:
Pack of 4
Product Code. 11894471
Brand: Thermo Scientific™ 75003742
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
  • Maximum tube size: 63 × 130mm
TRUSTED_SUSTAINABILITY
Specifications
Type Swinging Bucket Adapter
Capacity 100 mL Round-bottom Open-top Tube