missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXK, Mouse anti-Human, Clone: 1H5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007294-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: LIPSNYVTENKITNLEIYEWYHRNITRNQAEHLLRQESKEGAFIVRDSRHLGSYTISVFMGARRSTEAAIKHYQIKKNDSGQWYVAERHAFQSIPELIWYSpecifications
| TXK | |
| Monoclonal | |
| Unconjugated | |
| TXK tyrosine kinase | |
| BTKL/MGC22473/PSCTK5/PTK4/RLK/TKL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1H5 | |
| In 1x PBS, pH 7.4 | |
| NM_003328 | |
| TXK | |
| TXK (NP_003319, 131 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7294 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction