missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TXK, GST Tag Recombinant Protein

Product Code. 10277564
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
Each
This item is not returnable. View return policy

Product Code. 10277564

Brand: Invitrogen™ PV5860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic Domain.

TXK, along with the close relatives BMS, ITK, and BTK, signal downstream of antigen receptors and other types of receptors. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain a pleckstrin homology (PH) and a Tec homology (TH) domain in their amino termini. Four alternatively spliced forms of Tec are found to be expressed broadly in cells of hematopoietic lineage and hepatocytes. The Emt gene product associates with CD28 and becomes activated subsequent to CD28 ligation. Btk is necessary for proper B cell development, and mutations in the gene encoding Btk have been associated with families suffering from X-linked agammaglobulinemia, also referred to as Bruton's disease. The Bmx protein shares a high degree of homology with Btk and seems to be expressed at highest levels in the heart. Txk expression is T cell-specific, while expression of the Drosophila Tec homolog, Dsrc28C, is developmentally regulated.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42681
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with no preservative
Gene ID (Entrez) 7294
Molecular Weight (g/mol) 58.5 kDa
Name Human TXK, GST Tag
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK
Common Name TXK
Gene Symbol TXK
KinaseFamily TEC Kinase Family
Protein Length 260-527
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 260-527
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity
Kinase Group Tyrosine Kinases
Shipping Condition Dry Ice
System Compatibility LanthaScreen™
Tag Position N-Terminal
Target Entry TXK
Target Gene TXK
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic Domain
Form Liquid
Protein Subtype Cytoplasmic Tyrosine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.