missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXLNG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551465
This item is not returnable.
View return policy
Description
Immunogen sequence: VSPAYCTQES REEIPGGEAR TDPPDGQQDS ECNRNKEKTL GKEVLLLMQA LNTLSTPEEK LAALCKKYAD LLEESRSVQK QMKILQKKQA QIVKEKVHLQ SEHSKAILAR SKLESLCREL Q Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
TXLNG gene ontology annotations related to this gene include regulation of bone mineralization; regulation of cell cycle process.
Specifications
| TXLNG | |
| Polyclonal | |
| Unconjugated | |
| TXLNG | |
| 4932441K18Rik; CXHXORF15; CXorf15; ELRG; environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; factor inhibiting ATF4-mediated transcription; FIAT; gamma-taxilin; lipopolysaccharide specific response-5 protein; lipopolysaccharide-responsive gene protein; Lipopolysaccharide-specific response protein 5; Lrg; Lrp; LSR5; taxilin gamma; TXLNG; TXLNGX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 302680, 353170, 55787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8BHN1, Q9NUQ3 | |
| TXLNG | |
| Recombinant protein corresponding to Human TXLNG. Recombinant protein control fragment (Product #RP-91989). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction