missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXNDC16 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5148316

Product Code. 30019834

  • € 368.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TXNDC16 Polyclonal Antibody for IHC (P)

TXNDC16 gene ontology annotations related to this gene include cell redox homeostasis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TXNDC16
Polyclonal
Unconjugated
TXNDC16
5730420B22Rik; C77647; ERp90; Kiaa1344; RGD1306755; thioredoxin domain containing 16; thioredoxin domain-containing protein 16; TXNDC16
Rabbit
Antigen affinity chromatography, Protein A
RUO
57544
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
100 μg/mL
PBS with 0.03% ProClin 300; pH 7
Q9P2K2
TXNDC16
E. coli-derived Human TXNDC16 fragment.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.