missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXNDC3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5139787

Product Code. 17884300

  • € 464.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TKAGFIIEAE HKTVLTEEQV VNFYSRIADQ RDFEEFVSFM TSGLSYILVV SQGSKHNPPS EETEPQTDTE PNERSEDQPE VEAQVTPGMM KNKQDSLQEY LE Highest antigen sequence indentity to the following orthologs: Mouse - 50%, Rat - 43%.

This gene encodes a protein with an N-terminal thioredoxin domain and three C-terminal nucleoside diphosphate kinase (NDK) domains, but the NDK domains are thought to be catalytically inactive. The sea urchin ortholog of this gene encodes a component of sperm outer dynein arms, and the protein is implicated in ciliary function. Mutations in this gene are implicated in primary ciliary dyskinesia type 6.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TXNDC3
Polyclonal
Unconjugated
NME8
1700056P15Rik; CILD6; epididymis secretory protein Li 99; HEL-S-99; NM23-H8; NME/NM23 family member 8; Nme8; spermatid-specific thioredoxin-2; spermatid-specific thioredoxin-2 protein; sperm-specific thioredoxin 2; SPTRX2; sptrx-2; thioredoxin domain containing 3 (spermatozoa); thioredoxin domain-containing protein 3; Txndc3
Rabbit
Antigen affinity chromatography
RUO
51314
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.8 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N427
NME8
Recombinant protein corresponding to Human NME8. Recombinant protein control fragment (Product #RP-108790).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.