missing translation for 'onlineSavingsMsg'
Learn More

TXNDC5 Mouse anti-Human, Clone: OTI2E6, TrueMAB™, Origene

Product Code. 17527152
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17527152 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17527152 Supplier Origene Technologies Supplier No. TA507281S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TXNDC5 Monoclonal antibody specifically detects TXNDC5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TXNDC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Txndc5
Gene Accession No. Q8NBS9
Gene Alias AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700
Host Species Mouse
Immunogen Full length human recombinant protein of human TXNDC5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81567
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.