Learn More
TXNDC5 Mouse anti-Human, Mouse, Clone: OTI1C3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF507262
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| TXNDC5 | |
| Monoclonal | |
| Unconjugated | |
| TXNDC5 | |
| ENDOPDI, ERP46, HCC-2, HCC2, PDIA15, STRF8, UNQ364 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 105245, 81567 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8NBS9, Q91W90 | |
| Txndc5 | |
| Full length human recombit protein of human TXNDC5 produced in HEK293T cell | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.