missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523485
This item is not returnable.
View return policy
Description
TXNIP Polyclonal Antibody for Western Blot
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.
Specifications
| TXNIP | |
| Polyclonal | |
| Unconjugated | |
| TXNIP | |
| 1200008J08Rik; AA682105; ARRDC6; EST01027; HHCPA78; hyperlipidemia 1; Hyplip1; Tbp-2; THIF; thioredoxin binding protein 2; thioredoxin binding protein-2; thioredoxin interacting protein; thioredoxin-binding protein 2; Thioredoxin-interacting protein; TXNIP; upregulated by 1,25-dihydroxyvitamin D-3; VDUP1; Vitamin D3 up-regulated protein 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10628, 117514, 56338 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q5M7W1, Q8BG60, Q9H3M7 | |
| TXNIP | |
| A portion of amino acids 100-150 of human TxNIP. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction