missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXNL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15779700 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15779700 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779700 Supplier Invitrogen™ Supplier No. PA551748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CLNESDEHGF DNCLRKDTTF LESDCDEQLL ITVAFNQPVK LYSMKFQGPD NGQGPKYVKI FINLPRSMDF EEAERSEPTQ ALELTEDDIK EDGIVPLRYV KFQNVNSVTI FVQSNQGEEE TTRISYFTFI GTPVQATNMN DFKRVVGK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Thioredoxins are a group of small redox active proteins possessing a conserved active site sequence. TrxL (Thioredoxin like protein 1) has 2 distinct domains: an N terminal domain, which is 43% identical to human TXN, and a C terminal domain, which shows no homology to other proteins in the sequence databases. The active site sequence, located within the N terminal domain, is that of a thioredoxin like protein; compared to the active site sequence of TXN, it has a single amino acid substitution. Unlike other thioredoxin like proteins, TrxL does not serve as a substrate for thioredoxin reductase in an insulin assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Txnl1
Gene Accession No. O43396, Q8CDN6, Q920J4
Gene Alias 32 kDa thioredoxin-related protein; 32kDa; epididymis secretory protein Li 114; HEL-S-114; thioredoxin like 1; thioredoxin-like (32kD); thioredoxin-like 1; thioredoxin-like protein 1; thioredoxin-related 32 kDa protein; thioredoxin-related protein; thioredoxin-related protein 1; Trp; TRP32; TXL; TXL-1; Txnl; TXNL1
Gene Symbols Txnl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TXNL1. Recombinant protein control fragment (Product #RP-101856).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140922, 53382, 9352
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.