missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNL5 Monoclonal Antibody (544710)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524245
This item is not returnable.
View return policy
Description
This antibody detects endogenous human TRP14 in Western blots. Reconstitute at 0.5 mg/mL in sterile PBS.
TXNL5 disulfide reductase. May participate in various redox reactions through the reversible oxidation of its active center dithiol to a disulfide and catalyze dithiol-disulfide exchange reactions. Modulates TNF-alpha signaling and NF-kappa-B activation. Has peroxidase activity and may contribute to the elimination of cellular hydrogen peroxide.
Specifications
| TXNL5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9BRA2 | |
| TXNDC17 | |
| E. coli-derived recombinant human TRP14 Ala2-Asp123. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 544710 | |
| Unconjugated | |
| TXNDC17 | |
| 14 kDa thioredoxin-related protein; 4831443O22Rik; D11Ertd672e; MGC14353; protein 42-9-9; putative 42-9-9 protein; testicular tissue protein Li 214; thioredoxin (Trx)-related protein, 14 kDa; thioredoxin domain containing 17; thioredoxin domain-containing protein 17; thioredoxin-like 5; thioredoxin-like protein 5; TRP14; Txndc17; Txnl5 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 84817 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction