missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNRD2 Antibody (PSH02-26), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33068
This item is not returnable.
View return policy
Description
TXNRD2 Monoclonal antibody specifically detects TXNRD2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TXNRD2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| EC 1.8.1, EC 1.8.1.9, KIAA1652, Selenoprotein Z, SelZ, thioredoxin reductase 2, thioredoxin reductase 3, thioredoxin reductase beta, Thioredoxin reductase TR3, TR, TR3, TR-BETA, TRXR2thioredoxin reductase 2, mitochondrial | |
| Recombinant protein within human TXNRD2 aa 1-524/ 524. (Uniprot: Q9NNW7) | |
| 100 μL | |
| Breast Cancer, Membrane Trafficking and Chaperones | |
| 10587 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| PSH02-26 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction