missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNRD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16360-1-AP-20UL
This item is not returnable.
View return policy
Description
Thioredoxin reductase (TR) is a dimeric NADPH-dependent FAD containing enzyme that catalyzes the reduction of the active site disulfide of thioredoxin and other substrates. TR is a member of a family of pyridine nucleotide-disulfide oxidoreductases and is a key enzyme in the regulation of the intracellular redox environment. Three thioredoxin reductase genes have been found that encode selenocysteine containing proteins. This gene partially overlaps the COMT gene on chromosome 22.Specifications
| TXNRD2 | |
| Polyclonal | |
| Unconjugated | |
| TXNRD2 | |
| KIAA1652, Selenoprotein Z, SELZ, thioredoxin reductase 2, Thioredoxin reductase TR3, TR, TR BETA, TR3, TRXR2, TXNRD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10587, 26462, 50551 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JLT4, Q9NNW7, Q9Z0J5 | |
| Txnrd2 | |
| TXNRD2 Fusion Protein Ag8367 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction