missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 AL TF630 WG
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 2
Description
- Ventilated coverall available in sizes small to 3X.
- Attached non-dissipative under gloves.
- Integrated dissipative socks with boot flap.
- Gray.
- The protective system made of Tychem™ 6000 AL TF630 WG and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates.
- The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
- The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip.
- In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
- Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate.
- They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Specifications
Specifications
| Color | Gray |
| For Use With (Application) | Protecting from Contaminents |
| Hood Type | Fully Encapsulated |
| Material | Tychem |
| Size | Large |
| Closure Type | Horizontal Zipper with Double Flap Hook and Loop |
| Disposable | Single-use |
| Gender | Unisex |
| Latex Free | Yes |
| No. of Pockets | None |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction