Learn More
Masterflex™ Tygon™ 2 Stop Hydrocarbon Tubing
Product Code. 11997865
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.64 mm
0.89 mm
1.02 mm
1.14 mm
1.3 mm
1.42 mm
1.5 mm
1.85 mm
2.06 mm
2.29 mm
Unit Size:
Pack of 12
Product Code. 11997865
Brand: Masterflex™ 9560634
Only null left
Add to basket
Add to basket
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 2 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant
Specifications
Color Code Yellow/Yellow
Diameter (Metric) Inner 1.42 mm
Material Tygon F4040
Overall Length (Metric) 381 mm
Thickness (Metric) 0.85 mm
Packaging Quantity 12 Pack