Learn More
Masterflex™ Tygon™ 3 Stop Hydrocarbon Tubing
Product Code. 11937955
Click to view available options
Diameter (Metric) Inner:
1.02 mm
1.42 mm
1.52 mm
1.85 mm
2.54 mm
2.79 mm
Color Code:
Green/Green/Green
Purple/Purple/Orange
Purple/Purple/White
White/White/White
Yellow/Yellow/Blue
Yellow/Yellow/Yellow
Unit Size:
Pack of 12
Product Code. 11937955
Brand: Masterflex™ 9560528
Only null left
Add to basket
Edge
Add to basket
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 3 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant
Specifications
Color Code White/White/White
Description Color-code: White-White
Diameter (Metric) Inner 1.02 mm
Material Tygon F4040
Overall Length (Metric) 40 cm
Thickness (Metric) 0.85 mm
For Use With (Application) For hydrocarbons, petroleum products and distillates. MS/CA cassettes.
Min. Operating Temperature (Metric) -37°C
Packaging Quantity 12 Pack
Tubing Size 1.02 mm I.D., 2.70 mm O.D.
Gas Permeability CO2 100, O2 22, N2 12
Limitations Not recommended for strong acids and alkalies, foodstuffs, beverages and medicines. Potential leaching of plasticizers.
Color Translucent Yellow
Max. Operating Temperature (Metric) 74°C
Product Type 3-stop Tubing