Learn More
Saint Gobain Life Sciences Tygon™ 2765 Mill Tubing
Click to view available options
Diameter (Metric) Inner:
1.14 mm
1.52 mm
2.79 mm
Color:
Blue, Yellow
Purple/White
Red
Unit Size:
Pack of 6
Description
- For use with highly aggressive chemicals
- Long service life
- High fatigue resistance due to flexing
Specifications
Specifications
| Certifications/Compliance | FDA Certified |
| Color Code | Red |
| Autoclavable | Autoclavable |
| Diameter (English) Inner | 0.04 in. |
| Diameter (Metric) Inner | 1.14 mm |
| Material | DEHP |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.85 mm |
| For Use With | Masterflex Peristaltic Pumps, Ismatec, Gilson, Watson-Marlow |
| Packaging Quantity | 6 Pk. |
| Show More |