Learn More
Masterflex™ Tygon™ Non-DEHP Microbore Tubing
Product Code. 15187044
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.76 mm
1.27 mm
Diameter (Metric) Outer:
0.76 mm
2.29 mm
Unit Size:
150m
Product Code. 15187044
Brand: Masterflex™ 0642003
Only null left
Add to basket
Add to basket
For use in intravenous or intra-arterial infusion, and other surgical and hospital applications. Saint Gobain Performance Plastics™ Tygon™ Non-DEHP Microbore Tubing is rigid enough for easy handling, yet soft enough to reduce problems with puncturing.
  • High rigidity for kink resistance
  • Meets USP Class VI criteria
  • Compatible with blood
  • Clear, with a glass-smooth inner bore
  • Reduced flexibility for suction applications
  • REACH compliant
  • Thermoplastic in nature
Specifications
Certifications/Compliance Resin meets USP Class VI requirements, non-DEHP
Autoclavable Autoclavable
Diameter (Metric) Inner 0.76 mm
Material Tygon
Diameter (Metric) Outer 2.29 mm
Overall Length (Metric) 152.4 m
Thickness (Metric) 0.76 mm
For Use With (Application) General laboratory and pharmaceutical use, especially with precision injection or dispensing applications
Min. Operating Temperature (Metric) -35°C
Packaging Quantity 1 Pack
Sterility Sterile
Color Translucent
Flexibility Flexible
Max. Operating Temperature (Metric) 85°C