Learn More
Heidolph™ Tygon (Standard) Tubing
Product Code. 10209561 Shop All Heidolph Instruments Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.7 mm
3.1 mm
7.9 mm
Unit Size:
1m
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10209561 1.7 mm Each
10085751 0.8 mm Each
10351632 7.9 mm 1m
10628582 3.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10209561 Supplier Heidolph™ Supplier No. 5256400000
Only null left
Add to basket
Add to basket
Suitable for standard applications. Heidolph™ Tygon™ Standard Tubing is durable and resistant.
  • Non-toxic.
  • Non-oxidizing.
  • Good resistance to acids, bases and inorganic media.
  • Very low gas permeability.
  • Good performance life.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance FDA (21 CFR 177.2601) and GLP
Autoclavable Autoclavable
Diameter (Metric) Inner 1.7 mm
Material Thermoplastic
Diameter (Metric) Outer 4.9 mm
Thickness (Metric) 1.6 mm
For Use With Pump
For Use With (Application) Non-Toxic, Non-Oxidizing, Good Resistance to Acids, Bases and Inorganic Media, Very Low Gas Permeability, Good Performance Life.
Min. Operating Temperature (Metric) -50°C
Sterility Non-sterile
Sterilization Method Autoclave for 30 min (at 1 bar and 120°C (material may change color) or with ethylene oxide.)
Height (Metric) 8.8 m Water
Gas Permeability Very low
Limitations Segregation of plasticizers is possible.
Color Transparent
Maximum Pressure Continuous/short time 0.7/1.7 bar
Vacuum Level Good
Max. Operating Temperature (Metric) 75°C
Product Type Tubing