missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYKi Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82045V
This item is not returnable.
View return policy
Description
TYKi Polyclonal antibody specifically detects TYKi in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| TYKi | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| DyLight 405 | |
| cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2 | |
| TYKi antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human TYKi. The immunogen is located within amino acids 370 - 420 of TYKi. Amino Acid Squence: PEERLQRLQGRGMEKTREE | |
| 0.1 mL | |
| Protein Kinase | |
| 129607 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction