missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYMP Monoclonal Antibody (OTI7A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800998S
This item is not returnable.
View return policy
Description
TYMP Monoclonal antibody specifically detects TYMP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TYMP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P19971 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1890 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7A6 | |
| Unconjugated | |
| TYMP | |
| 2900072D10Rik; ECGF; Ecgf1; endothelial cell growth factor 1 (platelet-derived); gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; thymidine phosphorylase; TP; TYMP | |
| Human recombinant protein fragment corresponding to amino acids 311-482 of human TYMP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction