missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYMS Monoclonal Antibody (OTI4G10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801793S
This item is not returnable.
View return policy
Description
TYMS Monoclonal antibody specifically detects TYMS in Human samples. It is validated for Western BlotSpecifications
| TYMS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04818 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7298 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4G10 | |
| Unconjugated | |
| TYMS | |
| HST422; MGC88736; OK/SW-cl.29; Thymidylate synthase; thymidylate synthetase; TMS; TS; Tsase; Tyms; tysy | |
| Full length human recombinant protein of human TYMS produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction