Learn More
TYMS Mouse anti-Human, Clone: OTI1C4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF801811
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Thymidylate Synthase (TS) (EC 2.1.1.45) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.Specifications
| TYMS | |
| Monoclonal | |
| Unconjugated | |
| TYMS | |
| HST422, TMS, TS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7298 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1C4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P04818 | |
| TYMS | |
| Full length human recombit protein of human TYMS produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.