missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet V-MOVE-90 Light Gray
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3503400136111
Additional Details : Weight : 86.16600kg
Alert:
For indoor use only
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawer opens and closes at the push of a button
- Perfect access: the vertical drawer provides an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawer lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1750 mm | |
4 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
775 mm | |
Steel | |
860 x 449 x 1966 mm | |
775 x 331 x 1750 mm | |
Tray shelf | |
1966 mm | |
V90.196.045.VDAC:0012 | |
1 |
Light gray vertical drawer doors | |
Safety storage cabinet | |
331 mm | |
Powder-coated plain | |
449 mm | |
395.50 kg | |
Light Gray | |
860 mm | |
Auto-close, Sliding | |
4 | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |