missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Antibody (SPM360), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34770JF549
This item is not returnable.
View return policy
Description
Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Tyrosinase | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
| Recombinant tyrosinase protein (Uniprot: P14679 ) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 7299 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| SPM360 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction