missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Antibody (TYR/2024R), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08892B
This item is not returnable.
View return policy
Description
Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Tyrosinase | |
| Monoclonal | |
| Biotin | |
| CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
| Recombinant human full-length tyrosinase (TYR) protein (Uniprot: P14679) | |
| 100 μg | |
| Lipid and Metabolism | |
| 7299 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| TYR/2024R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction