missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Antibody (TYR/3829), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08891JF549
This item is not returnable.
View return policy
Description
Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Tyrosinase | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
| Recombinant fragment (around aa332-474) of human Tyrosinase protein (exact sequence is proprietary) (Uniprot: P14679 ) | |
| 100 μg | |
| Lipid and Metabolism | |
| 7299 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| TYR/3829 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction