missing translation for 'onlineSavingsMsg'
Learn More

Tyrosine Hydroxylase Antibody (OTI1A12), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

€ 256.00 - € 550.00

Specifications

Antigen Tyrosine Hydroxylase
Clone OTI1A12
Dilution Western Blot 1:4000, Immunohistochemistry : 1:150, Immunocytochemistry/ Immunofluorescence 1:100
Applications Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306579
View Documents
Novus Biologicals
NBP3-44720-100ul
100 μL
€ 550.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306491
View Documents
Novus Biologicals
NBP3-44720-30ul
30 μL
€ 256.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Tyrosine Hydroxylase
Western Blot 1:4000, Immunohistochemistry : 1:150, Immunocytochemistry/ Immunofluorescence 1:100
Monoclonal
Purified
RUO
Human, Mouse, Rat
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Full length human recombinant protein of human TH(NP_000351) produced in HEK293T cell.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
OTI1A12
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Mouse
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer, Tyrosine Kinases
PBS (pH 7.3) 1% BSA, 50% glycerol
7054
IgG1
>95%
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.