missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosine Hydroxylase Antibody (OTI3H3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
€ 256.00 - € 550.00
Specifications
| Antigen | Tyrosine Hydroxylase |
|---|---|
| Clone | OTI3H3 |
| Dilution | Western Blot 1:400-4000, Immunocytochemistry/ Immunofluorescence 1:100 |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306440
|
Novus Biologicals
NBP3-44857-30ul |
30 μL |
€ 256.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306550
|
Novus Biologicals
NBP3-44857-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Tyrosine Hydroxylase | |
| Western Blot 1:400-4000, Immunocytochemistry/ Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
| Full length human recombinant protein of human TH(NP_000351) produced in HEK293T cell. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI3H3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer, Tyrosine Kinases | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 7054 | |
| IgG2a | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title