missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tyrosine Hydroxylase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118306
This item is not returnable.
View return policy
Description
Tyrosine Hydroxylase Polyclonal Antibody for Western Blot, ICC/IF, IP
Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.
Specifications
| Tyrosine Hydroxylase | |
| Polyclonal | |
| PBS with no preservative | |
| P07101 | |
| TH | |
| A synthetic peptide corresponding to the C-terminus of the Human Tyrosine Hydroxylase/TH. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| TH | |
| dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7054 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction