Learn More
Tyrosine kinase-IN-1, MedChemExpress
Product Code. 16450619
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16450619
Brand: Medchem Express HY100315
MedChemExpress Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC50s of 4, 20, 4, 2 nM for KDR, Flt-1, FGFR1 and PDGFRα, respectively.
Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC50s of 4, 20, 4, 2 nM for KDR, Flt-1, FGFR1 and PDGFRα, respectively.
Specifications
Chemical Name or Material Tyrosine kinase-IN-1
CAS 705946-27-6
Color Brown
Purity Grade Notes Research
Molecular Formula C26H28FN5O
Quantity 1 mg
Solubility Information DMSO : ≥ 62.5 mg/mL (140.28 mM)
SMILES O=C1NC2=C(C=C(NC3CCN(CC)CC3)C=C2)/C1=C(C4=CC=CC(F)=C4)/C5=NC=C(C)N5
Molecular Weight (g/mol) 445.53
Formula Weight 445.53
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only