missing translation for 'onlineSavingsMsg'
Learn More

Tyrosine kinase-IN-1, MedChemExpress

Product Code. 16445677
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16445677

Brand: Medchem Express HY100315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC50s of 4, 20, 4, 2 nM for KDR, Flt-1, FGFR1 and PDGFRα, respectively.

Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC50s of 4, 20, 4, 2 nM for KDR, Flt-1, FGFR1 and PDGFRα, respectively.

Specifications

Chemical Name or Material Tyrosine kinase-IN-1
CAS 705946-27-6
Color Brown
Purity Grade Notes Research
Molecular Formula C26H28FN5O
Quantity 5 mg
Solubility Information DMSO : ≥ 62.5 mg/mL (140.28 mM)
SMILES O=C1NC2=C(C=C(NC3CCN(CC)CC3)C=C2)/C1=C(C4=CC=CC(F)=C4)/C5=NC=C(C)N5
Molecular Weight (g/mol) 445.53
Formula Weight 445.53
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.