missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosylprotein Sulfotransferase 1/TPST1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93115-0.02ml
This item is not returnable.
View return policy
Description
Tyrosylprotein Sulfotransferase 1/TPST1 Polyclonal antibody specifically detects Tyrosylprotein Sulfotransferase 1/TPST1 in Rat samples. It is validated for Western Blot
Specifications
| Tyrosylprotein Sulfotransferase 1/TPST1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 2.8.2.20, protein-tyrosine sulfotransferase 1, tyrosylprotein sulfotransferase 1TPST-1, tyrosylprotein sulfotransferase-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-75 of human Tyrosylprotein Sulfotransferase 1/TPST1 (NP_003587.1). MVGKLKQNLLLACLVISSVTVFYLGQHAMECHHRIEERSQPVKLESTRTTVRTGLDLKANKTFAYHKDMPLIFIG | |
| 0.02 mL | |
| Cardiovascular Biology, Immunology | |
| 8460 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction