missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYRP1 Antibody (TA99) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34720-0.1mg
This item is not returnable.
View return policy
Description
TYRP1 Monoclonal antibody specifically detects TYRP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF
Specifications
| TYRP1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5-1 μg/mL, Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Frozen 0.5-1 μg/mL, Flow (Intracellular), CyTOF-ready | |
| b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
| SK-MEL-23 cells | |
| 0.1 mg | |
| Cancer | |
| 7306 | |
| Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF | |
| TA99 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction