missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TYRP1 Monoclonal Antibody (CL4923)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA535023
This item is not returnable.
View return policy
Description
Immunogen sequence: VCDICTDDLM GSRSNFDSTL ISPNSVFSQW RVVCDSLEDY DTLGTLCNST EDGPIRRNPA GNVARPMVQR LPEPQDVAQC LEVGLFDTPP FYSNSTNSFR NTVEGYSDPT GKYDPAVRSL H Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a melanosomal enzyme that belongs to the tyrosinase family and plays an important role in the melanin biosynthetic pathway. Defects in this gene are the cause of rufous oculocutaneous albinism and oculocutaneous albinism type III.
Specifications
| TYRP1 | |
| Monoclonal | |
| 1.00 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17643 | |
| TYRP1 | |
| Recombinant protein fragment of Human TYRP1 (Product #RP-96731) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL4923 | |
| Unconjugated | |
| TYRP1 | |
| 5,6-dihydroxyindole-2-carboxylic acid oxidase; B; b-PROTEIN; brown; Brown locus protein; CAS2; Catalase B; CATB; DHICA oxidase; glycoprotein 75; GP75; iris stromal atrophy; isa; melanogenic enzyme; melanoma antigen gp75; MGC120175; MGC149362; OCA3; RP11-3L8.1; TRP; TRP1; TRP-1; truncated Tyrp1; TRY1; TRY4; TRYP1; tyrosinase related protein 1; tyrosinase-related protein 1; tyrosinase-related protein 1 precursor; tyrosine phosphatase 1, same as B (Brown); TYRP; Tyrp1; Tyrp-1; TYRRP | |
| Mouse | |
| Protein A | |
| RUO | |
| 7306 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur