missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin AG 879, MedChemExpress
Description
Tyrphostin AG 879 (AG 879) is a tyrosine kinase inhibitor that inhibits TrKA phosphorylation (IC50 of 10 μM), but not TrKB and TrKC. Tyrphostin AG 879 is also a selective ErbB2 tyrosine kinase inhibitor with an IC50 of 1 μM, and has at least 500-fold higher selectivity to ErbB2 than EGFR. Tyrphostin AG 879 has anticancer activity.
Specifications
Specifications
| Chemical Name or Material | Tyrphostin AG 879 |
| CAS | 148741-30-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H24N2OS |
| Quantity | 10 mg |
| Synonym | AG 879 |
| Solubility Information | DMSO : 50 mg/mL (158.00 mM; Need ultrasonic) |
| SMILES | S=C(N)/C(C#N)=C/C1=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C1 |
| Molecular Weight (g/mol) | 316.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?