missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 879, MedChemExpress

Product Code. 16443736
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443736 25 mg 25mg
16478018 10 mg 10mg
16470478 5 mg 5mg
16413947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443736 Supplier Medchem Express Supplier No. HY20878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tyrphostin AG 879 (AG 879) is a tyrosine kinase inhibitor that inhibits TrKA phosphorylation (IC50 of 10 μM), but not TrKB and TrKC. Tyrphostin AG 879 is also a selective ErbB2 tyrosine kinase inhibitor with an IC50 of 1 μM, and has at least 500-fold higher selectivity to ErbB2 than EGFR. Tyrphostin AG 879 has anticancer activity.

Tyrphostin AG 879 (AG 879) is a tyrosine kinase inhibitor that inhibits TrKA phosphorylation (IC50 of 10 μM), but not TrKB and TrKC. Tyrphostin AG 879 is also a selective ErbB2 tyrosine kinase inhibitor with an IC50 of 1 μM, and has at least 500-fold higher selectivity to ErbB2 than EGFR. Tyrphostin AG 879 has anticancer activity.

Specifications

Chemical Name or Material Tyrphostin AG 879
CAS 148741-30-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H24N2OS
Quantity 25 mg
Synonym AG 879
Solubility Information DMSO : 50 mg/mL (158.00 mM; Need ultrasonic)
SMILES S=C(N)/C(C#N)=C/C1=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C1
Molecular Weight (g/mol) 316.46
Formula Weight 316.46
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.